Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

PBKH9AA74667

PharmaBlock (USA), Inc.

3,3-dimethyl-2H-pyrazolo[5,1-b]oxazole-7-carboxylic acid

Synonym(s):

3,3-dimethyl-2,3-dihydropyrazolo[5,1-b]oxazole-7-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Product PBKH9AA74667 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C8H10O3N2
CAS Number:
Molecular Weight:
182.18
MDL number:

Skip To

Inchi Code

1S/C8H10N2O3/c1-8(2)4-13-6-5(7(11)12)3-9-10(6)8/h3H,4H2,1-2H3,(H,11,12)

Inchi Key

PHKZBNMEWGJGFB-UHFFFAOYSA-N

Purity

97%

Storage temp.

Refrigerated storage

Country of Origin

CN

Physical Form

Solid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library