MilliporeSigma
All Photos(1)

11420470001

Roche

dUTP

=98%, solution, 100 mM, suitable for DNA amplification

Synonym(s):
2′-deoxy-uridine-5′-triphosphate, deoxy-uridine-5′-triphosphate, 2′-

description

Lithium salt

Assay

98%

form

solution

usage

sufficient for 6,250 standard reactions (PCR)

packaging

pkg of 250 μL (25 μmol; 100 mM)

manufacturer/tradename

Roche

concentration

100 mM

technique(s)

DNA amplification: suitable
DNA sequencing: suitable

storage temp.

−20°C

General description

dUTP is a 100 mM clear, colorless solution of the lithium salt (pH 7).

Contents
dUTP solution in water (pH 7), 100 mM, lithium salt

Application

DNA that contains dUTP is susceptible to hydrolysis with Uracil-DNA Glycosylase. Therefore, dUTP is most commonly used (e.g., in PCRs and RT-PCRs with Taq DNA Polymerase) to prevent carryover contamination.However, dUTP is also a substrate for other DNA synthesizing enzymes (e.g., the Kornberg DNA polymerase and the Klenow Enzyme). Therefore, dUTP can be used for DNA sequencing, labeling, and all types of amplification reactions or primer-extension reactions.

Quality

Typical analysis:
  • ≥98% dUTP (HPLC), =1.5% dUDP (HPLC), =0.5% dUMP (HPLC)
  • Absorbance determination A250/A260, A280/A260, A290/A260

Unit Definition

1 μmol dUTP = 9.9 A260 units

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 2

1 of 2

Biotin-16-dUTP

Roche

11093070910

Biotin-16-dUTP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service