MilliporeSigma
All Photos(1)

KLEN-RO

Roche

Klenow Enzyme

Labeling grade, from Escherichia coli lysogenic NM 964

Synonym(s):
Klenow
Enzyme Commission number:

biological source

Escherichia coli

Quality Level

form

solution

specific activity

>5000 units/mg protein

packaging

pkg of 100 U (11008404001)
pkg of 500 U (11008412001)

manufacturer/tradename

Roche

storage temp.

−20°C

General description

Klenow enzyme is a DNA-dependent 5′→3′ polymerase with 3′→5′ exonuclease activity. It lacks the 5′→3′ exonuclease activity of the native enzyme. The addition of mononucleotides from dNTPs to the 3′-OH terminus of DNA is catalyzed. This property is used to synthesize DNA complementary to single-stranded DNA templates.

Specificity

Heat inactivation: Add 2 μl 0.2 mM EDTA (pH 8.0) and/or heat to 65 °C for 10 minutes

Packaging

100, 500 units

Application

Use Klenow Enzyme for:
  • Random-primed DNA labeling using random oligonucleotides as primers for the incorporation of nonradioactively labeled and 32P-labeled nucleotides.
  • Fill-in reaction for blunt-end formation of 3′-recessed (staggered) ends.

Quality

Absence of endo- and exonuclease activity tested according to the current Quality Control procedures

Unit Definition

One unit is the enzyme activity which incorporates 10 nmol of total nucleotides into an acid-precipitable fraction in 30 minutes under assay conditions at +37 °C with poly [d(A-T)] as primer (Richardson, C.C. & Kornberg, A. (1994) J. Biol. Chem. 244, 2996).

Volume Activity: 2,000 U/ml

Physical form

Enzyme in Storage buffer: 50mM potassium phosphate, 1mM dithioerythritol, 50% glycerol (v/v), pH 7.0 (+4°C).

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(C)

does not flash

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Product Information Sheet

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service