Sign In to View Organizational & Contract Pricing.
Select a Size
Pricing and availability is not currently available.
About This Item
Linear Formula:
NaH2PO2 · H2O
CAS Number:
Molecular Weight:
105.99
UNSPSC Code:
12352106
PubChem Substance ID:
EC Number:
231-669-9
MDL number:
Assay:
80.0-86.0% anhydrous basis
Grade:
SAJ first grade
Form:
solid
Skip To
Product Name
Sodium hypophosphite monohydrate, SAJ first grade, 80.0-86.0% anhydrous basis
InChI key
PLZNPHDJGFDNRM-UHFFFAOYSA-M
InChI
1S/Na.H3O2P.H2O/c;1-3-2;/h;3H2,(H,1,2);1H2/q+1;;/p-1
SMILES string
O.[Na+].[O-][PH2]=O
grade
SAJ first grade
assay
80.0-86.0% anhydrous basis
form
solid
availability
available only in Japan
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lukas J Goossen et al.
Chemical communications (Cambridge, England), (8)(8), 836-837 (2002-07-19)
A catalyst generated in situ from palladium acetate and tricyclohexylphosphine efficiently catalyzes the reduction of carboxylic acids with sodium hypophosphite in the presence of pivalic anhydride to give aldehydes with high selectivity. The low cost and convenient handling of the
Amy K Manocchi et al.
Langmuir : the ACS journal of surfaces and colloids, 27(11), 7052-7058 (2011-04-28)
We present an examination of palladium (Pd) nanoparticle growth on genetically modified tobacco mosaic virus (TMV1cys) nanotemplates via in situ small-angle X-ray scattering (SAXS). Specifically, we examine the role of the TMV1cys templates in Pd nanoparticle formation through the electroless
R Wadhwa et al.
Gerontology, 35(1), 14-18 (1989-01-01)
Addition of antioxidant, sodium hypophosphite (SHP; 1 mM), to the diet of Zaprionus paravittiger (Diptera) results in depressed ATPase activity in both the sexes in comparison to the normal group. However, the rate of age-related decline in ATPase activity is
W S Price et al.
Biophysical chemistry, 33(3), 205-215 (1989-07-01)
A 31P-NMR method, which complements earlier 13C-NMR procedures for probing the intra-erythrocyte microenvironment, is described. Hypophosphite is an almost unique probe of the erythrocyte microenvironment, since it is rapidly transported into the cell via the band 3 protein, and intra-
C Sánchez-Pedreño et al.
The Analyst, 115(9), 1257-1260 (1990-09-01)
A kinetic method for the determination of Te based on its inhibitory effect on the PdII-catalysed reaction between pyronine G and H2PO2- is described. The influence of experimental variables on the rate of the process and the potential interfering effect
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service