01070

Sigma-Aldrich

2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione

≥98.0% (HPLC)

Synonym(s):
2-Acetyldimedone, Dde
Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
Beilstein/REAXYS Number:
2089698
MDL number:
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (HPLC)

application(s)

peptide synthesis: suitable

mp

36-40 °C

SMILES string

CC(=O)C1C(=O)CC(C)(C)CC1=O

InChI

1S/C10H14O3/c1-6(11)9-7(12)4-10(2,3)5-8(9)13/h9H,4-5H2,1-3H3

InChI key

ITSKWKZDPHAQNK-UHFFFAOYSA-N

Other Notes

Protecting group reagent for primary amines; cleavage occurs orthogonally to other amine protections

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

M Gu et al.
Journal of dairy science, 100(10), 8265-8276 (2017-08-07)
Buffalo DGAT1 (diacylglycerol O-acyltransferase 1) was mainly investigated for the characterization of the gene itself and for the identification of the K232A polymorphism, similar to what has been accomplished in cattle, although no information has been reported so far at...
B García-Béjar et al.
Journal of applied microbiology, 130(3), 650-664 (2020-07-30)
This study aims to identify the yeast species and strains which entitled an unstudied area of Spain and evaluate the yeast species diversity richness and the genetic variety. A total of 702 yeasts were isolated from different environments in a...
Simone Cristina Pinto Matheus Fischer et al.
Arquivos brasileiros de cardiologia, 110(1), 16-23 (2018-02-08)
Metabolic syndrome (MS) is a condition that, when associated with ischemic heart disease and cardiovascular events, can be influenced by genetic variants and determine more severe coronary atherosclerosis. To examine the contribution of genetic polymorphisms to the extension and severity...
Xin-Jie Zhu et al.
American journal of rhinology & allergy, 34(5), 597-603 (2020-03-29)
Polymorphism -509C/T in the promoter of transforming growth factor beta1 (TGFB1) gene is implicated in the pathogenesis of asthma. This polymorphism might also act to regulate the development of allergic rhinitis (AR). To investigate whether -509C/T is associated with AR susceptibility...
In Wook Hwang et al.
Gene, 630, 8-12 (2017-08-10)
Mitochondria are subcellular organelles that contribute to aerobic ATP generation by oxidative phosphorylation (OXPHOS). Previous studies reported that mitochondrial dysfunction and deficiency caused by mitochondrial DNA polymorphisms is associated with various diseases. Especially, mitochondrial DNA 10398 A/G polymorphism is known...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.