Documents

03206

Sigma-Aldrich

Hydriodic acid

≥57%

Synonym(s):
Hydriotic acid
Empirical Formula (Hill Notation):
HI
CAS Number:
Molecular Weight:
127.91
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥57%

contains

hypophosphorous acid as stabilizer

impurities

≤0.001% heavy metals (as Pb)
≤0.025% iodine (I)
≤0.5% non-volatile matter

bp

127 °C (lit.)

anion traces

chloride, bromide (as Cl)-: ≤500 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Fe: ≤10 mg/kg

SMILES string

I

InChI

1S/HI/h1H

InChI key

XMBWDFGMSWQBCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 1787 8 / PGII

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Anthony Bertucci et al.
Bioorganic & medicinal chemistry letters, 21(2), 710-714 (2011-01-07)
The catalytic activity and the inhibition of a new coral carbonic anhydrase (CA, EC 4.2.1.1), from the scleractinian coral Stylophora pistillata, STPCA-2, has been investigated. STPCA-2 has high catalytic activity for the physiological reaction being less sensitive to anion and...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.