All Photos(1)

04055

Millipore

Gelatin from porcine skin

powder, suitable for microbiology

CAS Number:
EC Number:
MDL number:
NACRES:
NA.85

biological source

Porcine

Quality Level

sterility

non-sterile

form

powder

impurities

≤0.005% heavy metals (as Pb)
≤0.01% peroxides (as H2O2)
≤0.05% sulfur dioxide (SO2)

ign. residue

≤2%

loss

≤15% loss on drying

pH

3.8-7.6 (50 °C, 67 mg/mL in H2O)

gel strength

170-190 g Bloom67 mg/ml water

cation traces

As: ≤1 mg/kg

application(s)

microbiology

Looking for similar products? Visit Product Comparison Guide

Application

This product is recommended for use as a cell culture substratum at 1-5 μg/cm2 or 0.5-50 μg/mL. The optimal concentration does depend on cell type as well as the application and research objectives.

Gelatin has been used in many applications. It has use in coating cell culture to improve attachment of cells, being added to PCR to stabilize Taq DNA, as a blocking reagent in Western blotting, ELISA, and immunochemistry, and as a component of media for species differentiation in bacteriology. As a biocompatible polymer, it has used as a delivery vehicle for release of active biomolecules and in generation of scaffolds for tissue engineering applications. In the pharmaceutical industry, geltan can be used as a suspending and encapsulating agent, among other applications.

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Caution

Dry gelatin, when stored in airtight containers at room temperature, will remain unchanged for many years. When heated at 100°C in the presence of air, it swells becomes soft and disintegrates to a carbonaceous mass with evolution of pyridine bases and ammonia.

Preparation Note

This product is derived from porcine skin. Gelatin is soluble in hot than in cold water. It is practically insoluble in most organic solvents such as alcohol, chloroform, carbon disulfide, carbon tetrachloride, ether, benzene, acetone, and oils. The Bloom number, determined by the Bloom gelometer, is an indication of the strength of a gel formed from a solution of the known concentration. The Bloom number is proportional to the average molecular mass. Bloom numbers of porcine skin Gelatin vary from 90 to 300 g.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Xingshuo Zhang et al.
International journal of molecular sciences, 21(24) (2020-12-17)
Lower back pain is a leading cause of disability worldwide. The recovery of nucleus pulposus (NP) progenitor cells (NPPCs) from the intervertebral disc (IVD) holds high promise for future cell therapy. NPPCs are positive for the angiopoietin-1 receptor (Tie2) and
Leslie Crews et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(16), 4250-4260 (2008-04-18)
Altered expression and mutations in alpha-synuclein (alpha-syn) have been linked to Parkinson's disease (PD) and related disorders. The neurological alterations in PD patients have been associated with degeneration of dopaminergic cells and other neuronal populations. Moreover, recent studies in murine
Panuphong Pootawang et al.
Journal of nanoscience and nanotechnology, 13(1), 589-592 (2013-05-08)
The environmental concern pays much attention to the recent cause of the global warming effect. The reduction of the chemical uses is one of many ways to avoid this crucial problem. Herein, the green process for silver nanometallic particle formation
Kimberly E Hawkins et al.
Molecular brain, 6, 14-14 (2013-03-26)
Matrix metalloproteinases are important factors in the molecular mechanisms leading to neuronal injury in many neurological disorders. Matrix metalloproteinase (MMP)-9 is up-regulated after cerebral ischemia and neuroinflammation and is actively involved in blood-brain barrier disruption. Current methods of measuring MMP-9
Johno Breeze et al.
Journal of the Royal Army Medical Corps, 159(2), 84-89 (2013-05-31)
Ballistic gelatin is the most common tissue simulant used to reproduce the penetration of projectiles into muscle but published data to support its use are primarily based on bullets, despite explosive fragments being the most common cause of injury to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service