05729

Millipore

Agar

for chromogenic media

Synonym(s):
Agar-agar, Gum agar
Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
NACRES:
NA.85

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

microbiology: suitable (preparation)

ign. residue

≤4.0%

loss

≤10% loss on drying

turbidity

≤10 NTU

pH

6.5-6.8 (1.5% in gel, after autoclaving)

mp

≥85 °C

transition temp

gel point 35-36 °C

gel strength

≥750 g/cm2

anion traces

sulfate (SO42-): ≤2%

cation traces

Ca: ≤4000 mg/kg
Mg: ≤1200 mg/kg

Featured Industry

Food and Beverages

Looking for similar products? Visit Product Comparison Guide

Analysis Note

toxic substances: absence

storage_class_code

11 - Combustible Solids

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Arnaud Dechesne et al.
Applied and environmental microbiology, 74(16), 5195-5200 (2008-07-01)
Water is arguably the most important constituent of microbial microhabitats due to its control of physical and physiological processes critical to microbial activity. In natural environments, bacteria often live on unsaturated surfaces, in thin (micrometric) liquid films. Nevertheless, no experimental...
Queeny Wing-Han Yuen et al.
The open biomedical engineering journal, 5, 39-46 (2011-06-07)
Cervical lymph nodes are common sites of metastatic involvement in head and neck cancers. These lymph nodes are superficially located and palpation is a common practice for assessing nodal hardness and staging cancer which is, however, too subjective and with...
Malika Gouali et al.
Journal of clinical microbiology, 51(3), 894-900 (2013-01-04)
The performance of CHROMagar STEC and CHROMagar STEC O104 (CHROMagar Microbiology, Paris, France) media for the detection of Shiga toxin-producing Escherichia coli (STEC) was assessed with 329 stool specimens collected over 14 months from patients with suspected STEC infections (June...
Timothy A Wencewicz et al.
Journal of medicinal chemistry, 56(10), 4044-4052 (2013-04-26)
Chemical syntheses and biological evaluation of biscatecholate-monohydroxamate mixed ligand sideromycins utilizing the carbacephalosporin β-lactam antibiotic loracarbef and the fluoroquinolone antibiotic ciprofloxacin are described. The mixed ligand β-lactam sideromycin (1b) had remarkably selective and extremely potent antibacterial activity against the Gram-negative...
Extracellular bioluminescence of mycelium metabolites of the luminous mushroom Panellus stipticus (IBSO-2301) growing on an Agar medium.
A P Puzyr et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 448, 39-40 (2013-03-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service