07526

Supelco

4-[(9-Anthracenylmethylene)amino]-1,2-dihydro-1,5-dimethyl-2-phenyl-3H-pyrazol-3-one

suitable for fluorescence

Empirical Formula (Hill Notation):
C26H21N3O
CAS Number:
Molecular Weight:
391.46
Beilstein/REAXYS Number:
23002824
MDL number:
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

assay

≥98.0% (HPLC)

Quality Level

suitability

suitable for fluorescence

SMILES string

O=C(C(/N=C/C1=C(C=CC=C2)C2=CC3=C1C=CC=C3)=C(C)N4C)N4C5=CC=CC=C5

InChI

1S/C26H21N3O/c1-18-25(26(30)29(28(18)2)21-12-4-3-5-13-21)27-17-24-22-14-8-6-10-19(22)16-20-11-7-9-15-23(20)24/h3-17H,1-2H3/b27-17+

InChI key

RNXZNZGMBZVMDC-WPWMEQJKSA-N

Application

Highly selective fluorescent probe for Fe3+.
No interference from a large list of other ions, such as Ag+, Pb2+, Na+, K+, Hg2+, Cd2+, Ba2+, Zn2+, Cu2+, Ni2+, Ca2+, Al3+, Fe2+, SCN-, ....
Fluorescence data from resulting Fe3+ complex: excitation: 375 nm; emission 516 nm

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Yanmei Zhou et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 98, 14-17 (2012-09-18)
A novel and simple Schiff base composed with 9-anthraldehyde and 4-aminoantipyrine was synthesized and characterized as a fluorescent probe. In the presence of Fe(3+), the fluorescent intensity has a dramatic enhancement over other examined metal ions in aqueous solution. The...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.