11357

Supelco

1-Naphthyl isocyanate

for HPLC derivatization, LiChropur, ≥99.0% (GC)

Linear Formula:
C10H7NCO
CAS Number:
Molecular Weight:
169.18
Beilstein/REAXYS Number:
742779
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

grade

for HPLC derivatization

assay

≥99.0% (GC)

quality

LiChropur

application(s)

HPLC: suitable

refractive index

n20/D 1.631-1.635
n20/D 1.6344 (lit.)

bp

267 °C/761 mmHg (lit.)

mp

4 °C (lit.)

density

1.177 g/mL at 25 °C (lit.)

SMILES string

O=C=Nc1cccc2ccccc12

InChI

1S/C11H7NO/c13-8-12-11-7-3-5-9-4-1-2-6-10(9)11/h1-7H

InChI key

BDQNKCYCTYYMAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Target Organs

Respiratory system

RIDADR

UN 2206PSN1B 6.1 / PGIII

WGK Germany

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.