12510

Sigma-Aldrich

Benzoin

purum, ≥99.0% (GC)

Synonym(s):
α-Hydroxybenzyl phenyl ketone, α-Hydroxy-α-phenylacetophenone, 2-Hydroxy-2-phenylacetophenone
Linear Formula:
C6H5CH(OH)COC6H5
CAS Number:
Molecular Weight:
212.24
Beilstein/REAXYS Number:
391839
EC Number:
MDL number:
Pricing and availability is not currently available.

grade

purum

assay

≥99.0% (GC)

bp

194 °C/12 mmHg (lit.)

mp

133-135 °C
134-138 °C (lit.)

SMILES string

OC(c1ccccc1)C(=O)c2ccccc2

InChI

1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H

InChI key

ISAOCJYIOMOJEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

2

Flash Point(F)

338 °F

Flash Point(C)

170 °C

Javier Izquierdo et al.
Angewandte Chemie (International ed. in English), 51(47), 11686-11698 (2012-10-18)
N-Heterocyclic carbene (NHC) catalyzed transformations have emerged as powerful tactics for the construction of complex molecules. Since Stetter's report in 1975 of the total synthesis of cis-jasmon and dihydrojasmon by using carbene catalysis, the use of NHCs in total synthesis...
Tsuneomi Kawasaki et al.
Angewandte Chemie (International ed. in English), 50(35), 8131-8133 (2011-07-16)
Trigger happy: chiral oxygen isotopomers of hydrobenzoin ([(18)O](R)-1 and [(18)O](S)-1) acted as chiral triggers to induce the enantioselective addition of iPr(2)Zn to pyrimidine-5-carbaldehyde. An extremely small chiral influence arising from the presence of the oxygen isotope ((18)O) is amplified through...
Structures and reactivities of O-methylated Breslow intermediates.
Biplab Maji et al.
Angewandte Chemie (International ed. in English), 51(41), 10408-10412 (2012-09-13)
Qiu-Fen Hu et al.
Journal of natural products, 76(10), 1854-1859 (2013-09-26)
Eight new C-4-alkylated deoxybenzoins (1-8), three new diphenylethylenes (9-11), and five known diphenylethylenes were isolated from Arundina graminifolia. The structures of 1-11 were elucidated by spectroscopic methods including extensive 1D and 2D NMR techniques. Compounds 9-11 are the first naturally...
Umpolung by N-heterocyclic carbenes: generation and reactivity of the elusive 2,2-diamino enols (Breslow intermediates).
Albrecht Berkessel et al.
Angewandte Chemie (International ed. in English), 51(49), 12370-12374 (2012-10-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.