Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12625

Sigma-Aldrich

Aluminum potassium sulfate

puriss., anhydrous, calcined, ≥98% (calc with ref. to anhyd. subst.), powder

Synonym(s):

Potassium alum, Potassium aluminum sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KAl(SO4)2
CAS Number:
Molecular Weight:
258.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

puriss.

assay

≥98% (calc with ref. to anhyd. subst.)

form

powder

quality

calcined

impurities

≤0.005% heavy metals (as Pb)
≤0.05% ammonium (NH4)

loss

≤5% loss on drying, 250 °C

cation traces

As: ≤5 mg/kg
Fe: ≤100 mg/kg

SMILES string

[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.K.2H2O4S/c;;2*1-5(2,3)4/h;;2*(H2,1,2,3,4)/q+3;+1;;/p-4

InChI key

GRLPQNLYRHEGIJ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service