Skip to Content
MilliporeSigma

13703

Calcium silicate

purum, 12-22% Ca (as CaO) basis, 60-80% Si (as SiO2) basis, powder

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
UNSPSC Code:
12352302
PubChem Substance ID:
EC Number:
215-710-8
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Calcium silicate, purum, 12-22% Ca (as CaO) basis, 60-80% Si (as SiO2) basis, powder

InChI key

OYACROKNLOSFPA-UHFFFAOYSA-N

InChI

1S/Ca.O3Si/c;1-4(2)3/q+2;-2

SMILES string

[Ca++].[O-][Si]([O-])=O

grade

purum

form

powder

composition

Ca (as CaO), 12-22%
Si (as SiO2), 60-80%

loss

12-25% loss on ignition, 800 °C

density

2.9 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Salek et al.
Applied microbiology and biotechnology, 97(6), 2743-2752 (2012-11-29)
Silicate minerals can be a source of calcium and alkalinity, enabling CO2 sequestration in the form of carbonates. For this to occur, the mineral needs to be first dissolved in an acidifying process such as the biological process of anaerobic
Emma C Goethals et al.
Langmuir : the ACS journal of surfaces and colloids, 29(2), 658-666 (2012-10-20)
Polymer nanocapsules have gained an important place as drug delivery vehicles for a myriad of biomedical applications. However, the influence of nanocapsule size, wall thickness, and porosity toward controlling the drug delivery efficiency of nanocapsular systems is not well understood.
Bobo Yu et al.
Langmuir : the ACS journal of surfaces and colloids, 28(50), 17465-17476 (2012-11-23)
The aim was to determine the most effective calcium precursor for synthesis of sol-gel hybrids and for improving homogeneity of sol-gel bioactive glasses. Sol-gel derived bioactive calcium silicate glasses are one of the most promising materials for bone regeneration. Inorganic/organic
Yuandong Dou et al.
Acta biomaterialia, 8(11), 4139-4150 (2012-07-21)
How to accurately control the microstructure of bioactive inorganic/organic nanocomposites still remains a significant challenge, which is of great importance in influencing their mechanical strength and biological properties. In this study, using a combined method of electrospinning and hot press
Nana Wang et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 2294-2301 (2013-03-19)
A key challenge in tissue engineering is the construction of a scaffold with adequate properties which would mimic extracellular matrix (ECM) to induce the cells' efficient adhesion, proliferation and proper differentiation. Novel β-Ca2SiO4/PHBV composite scaffolds were fabricated by integrating β-Ca2SiO4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service