14436

Sigma-Aldrich

Zinc nitrate hexahydrate

purum, ≥98%

Linear Formula:
Zn(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
297.49
EC Number:
MDL number:
PubChem Substance ID:

grade

purum

assay

≥98%

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

Fe: ≤20 mg/kg
Pb: ≤100 mg/kg

SMILES string

O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.6H2O.Zn/c2*2-1(3)4;;;;;;;/h;;6*1H2;/q2*-1;;;;;;;+2

InChI key

JGPSMWXKRPZZRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 1514 5.1 / PGII

WGK Germany

3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.