16679

Supelco

Cuminaldehyde

analytical standard

Synonym(s):
4-Isopropylbenzaldehyde
Linear Formula:
(CH3)2CHC6H4CHO
CAS Number:
Molecular Weight:
148.20
Beilstein/REAXYS Number:
636547
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

assay

≥97.0% (GC)

form

neat

shelf life

limited shelf life, expiry date on the label

application(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.529 (lit.)
n20/D 1.531

bp

235-236 °C (lit.)

density

0.981 g/mL at 20 °C
0.977 g/mL at 25 °C (lit.)

Featured Industry

Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care

format

neat

SMILES string

[H]C(=O)c1ccc(cc1)C(C)C

InChI

1S/C10H12O/c1-8(2)10-5-3-9(7-11)4-6-10/h3-8H,1-2H3

InChI key

WTWBUQJHJGUZCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cuminaldehyde is a constituent of cumin oil that exhibits strong antifungal and antibacterial activities.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

hazcat

Acute Tox. 4 Oral

storage_class_code

10 - Combustible liquids

WGK Germany

WGK 2

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C

Certificate of Analysis

Certificate of Origin

A quantitative structure?antifungal activity relationship study of oxygenated aromatic essential oil compounds using data structuring and PLS regression analysis
Voda K, et al.
Journal of Molecular Modeling, 10(1), 76-84 (2004)
Seed Spices: Production, Quality, Export (2001)
Valério Monteiro-Neto et al.
PloS one, 15(5), e0232987-e0232987 (2020-05-15)
Escherichia coli and Staphylococcus aureus are important agents of urinary tract infections that can often evolve to severe infections. The rise of antibiotic-resistant strains has driven the search for novel therapies to replace the use or act as adjuvants of...
Natural Food Flavors and Colorants (2017)
Annemieke Strijkstra et al.
Applied and environmental microbiology, 80(24), 7592-7603 (2014-09-28)
The betaproteobacteria "Aromatoleum aromaticum" pCyN1 and "Thauera" sp. strain pCyN2 anaerobically degrade the plant-derived aromatic hydrocarbon p-cymene (4-isopropyltoluene) under nitrate-reducing conditions. Metabolite analysis of p-cymene-adapted "A. aromaticum" pCyN1 cells demonstrated the specific formation of 4-isopropylbenzyl alcohol and 4-isopropylbenzaldehyde, whereas with...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service