20892

Supelco

Cadmium standard for IC

ready-to-use, traceable to BAM, in nitric acid

Synonym(s):
Cadmium nitrate solution
CAS Number:
Beilstein/REAXYS Number:
8137359
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

concentration

in nitric acid
1.000 g/L

application(s)

ion chromatography: suitable

pH

≥3

format

single component solution

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

filtered through a 0.45 μm membrane filter

Preparation Note

prepared with Cd(NO3)2.4H2O and 0.001 M HNO3

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3082 9 / PGIII

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Sarita Nazareth et al.
Journal of basic microbiology, 48(5), 363-369 (2008-09-02)
Four different morphotypes of halotolerant Penicillium isolated from mangroves and salterns, selected on the basis of their morphological dissimilarities, namely, monoverticillate, biverticillate symmetric, biverticillate asymmetric and terverticillate, were studied for their response to the presence of heavy metals - lead...
Francesco Crea et al.
Annali di chimica, 95(11-12), 767-778 (2006-01-10)
In order to continue the investigation on the formation of hetero-metal polynuclear hydrolytic species, in this paper we report some results (at I = 0.16 mol L(-1) in NaNO3, at t = 25 degrees C by potentiometry, ISE-H+, glass electrode)...
Xue-Mei Li et al.
Journal of inorganic biochemistry, 101(8), 1165-1171 (2007-06-23)
A novel hybridization indicator, bis(benzimidazole)cadmium(II) dinitrate (Cd(bzim)(2)(NO(3))(2)), was utilized to develop an electrochemical DNA biosensor for the detection of a short DNA sequence related to the hepatitis B virus (HBV). The sensor relies on the immobilization and hybridization of the...
V S Fedenko et al.
Ukrains'kyi biokhimichnyi zhurnal (1999 ), 77(1), 104-109 (2005-12-13)
The effect of complexation between cyanidin (extracted from maize seedling roots) and cadmium ions was established in solution on the basis of absorption spectroscopy and colorimetry data. Spectral parameters for identification of the pigment added to the metal has been...
Akmal Muhammad et al.
Chemosphere, 60(4), 508-514 (2005-06-14)
A study was conducted to evaluate the effects of different concentrations of lead (Pb) and cadmium (Cd) applied as their nitrates on soil microbial biomass carbon (C(mic)) and nitrogen (N(mic)), and substrate utilization pattern of soil microbial communities. The C(mic)...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.