All Photos(1)

234095

Sigma-Aldrich

Methylene Blue zinc chloride double salt monohydrate

Synonym(s):
Basic Blue 9
Empirical Formula (Hill Notation):
C16H18ClN3S · 0.5ZnCl2 · H2O
CAS Number:
Molecular Weight:
406.02
Colour Index Number:
52015
Beilstein:
3839953
MDL number:
PubChem Substance ID:

λmax

664 nm

SMILES string

[Cl-].[H]O[H].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service