MilliporeSigma
All Photos(1)

270636

Supelco

Chloroform

suitable for HPLC, ≥99.8%

Synonym(s):
Methylidyne trichloride, Trichloromethane
Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
EC Number:
MDL number:
PubChem Substance ID:

vapor density

4.1 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

assay

≥99.8%

form

liquid

purified by

glass distillation

contains

amylenes as stabilizer

technique(s)

HPLC: suitable

impurities

Acetone and aldehyde, passes test
Acid and chloride, passes test
Free chlorine (Cl), passes test
H2SO4, passes test (darkened)
<0.010% water

evapn. residue

<0.0003%

color

APHA: ≤10

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

cation traces

Pb: ≤0.05 ppm

λ

H2O reference

UV absorption

λ: 245 nm Amax: 1.00
λ: 255 nm Amax: 0.150
λ: 260 nm Amax: 0.150
λ: 270 nm Amax: 0.020
λ: 290-400 nm Amax: 0.010

suitability

passes test for HPLC
passes test for dithizone test

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service