287652

Sigma-Aldrich

Cadmium chloride

ACS reagent, ≥99.0%

Synonym(s):
Cadmium dichloride, Dichlorocadmium
Empirical Formula (Hill Notation):
CdCl2
Molecular Weight:
183.32
EC Number:
MDL number:

grade

ACS reagent

vapor density

6.3 (vs air)

vapor pressure

10 mmHg ( 656 °C)

assay

≥99.0%

impurities

≤0.01% insolubles

mp

568 °C (lit.)

anion traces

NO3- and NO2-: ≤0.003%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.01%
Cu: ≤0.001%
Fe: ≤0.001%
K: ≤0.02%
NH4+: ≤0.01%
Na: ≤0.05%
Pb: ≤0.005%
Zn: ≤0.05%

SMILES string

Cl[Cd]Cl

InChI

1S/Cd.2ClH/h;2*1H/q+2;;/p-2

InChI key

YKYOUMDCQGMQQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

Certificate of Analysis

Certificate of Origin

Ana L Patrício Silva et al.
Environmental toxicology and chemistry, 34(9), 2159-2166 (2015-05-07)
Supralittoral ecosystems are among the most challenging environments for soil organisms, particularly when salinity fluctuations are involved, frequently combined with the presence of contaminants as a result of intense anthropogenic activities. Knowledge of how salinity influences the effect of contaminants...
Bart van der Burg et al.
Reproductive toxicology (Elmsford, N.Y.), 55, 95-103 (2014-12-22)
There is a great need for alternative testing methods for reproductive toxicants that are practical, fast, cost-effective and easy to interpret. Previously we followed a pragmatic approach using readily available tests, which was successful in predicting reproductive toxicity of chemicals...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service