MilliporeSigma
All Photos(1)

288586

Supelco

Silica gel

high-purity grade, pore size 60 Å, 2-25 μm particle size, without binder, with fluorescent indicator, pore volume 0.75 cm3/g, suitable for thin layer chromatography (TLC)

CAS Number:
EC Number:
MDL number:
PubChem Substance ID:

grade

high-purity grade

Quality Level

form

powder

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

impurities

≤0.01% Fe3+

particle size

2-25 μm

pore size

0.75 cm3/g pore volume
60 Å pore size

surface area

~500 m2/g

pH

~6.8 (10% in aq. suspension)

bp

2230 °C

mp

>1600 °C

anion traces

chloride (Cl-): ≤0.01%

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. It allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction

Packaging

1 kg in poly bottle

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service