About This Item
Skip To
grade
ACS reagent
puriss. p.a.
assay
≥99%
impurities
≤0.002% heavy metals (as Pb)
≤0.02% insoluble in HCl
anion traces
carbonate (CO32-): in accordance
chloride (Cl-): ≤20 mg/kg
chloride (Cl-): ≤50 mg/kg
nitrate (NO3-): ≤50 mg/kg
nitrate (NO3-): in accordance
cation traces
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤200 mg/kg
Na: ≤200 mg/kg
Sr: ≤500 mg/kg
SMILES string
O.O.[Ca++].[O-]S([O-])(=O)=O
InChI
1S/Ca.H2O4S.2H2O/c;1-5(2,3)4;;/h;(H2,1,2,3,4);2*1H2/q+2;;;/p-2
InChI key
PASHVRUKOFIRIK-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Other Notes
A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service