32231

Supelco

Chlorbenside sulfone

PESTANAL®, analytical standard

Synonym(s):
(4-Chlorobenzyl)-(4-chlorophenyl)sulfone, Chlorbensid-sulfone
Empirical Formula (Hill Notation):
C13H10Cl2O2S
CAS Number:
Molecular Weight:
301.19
Beilstein/REAXYS Number:
2130380
MDL number:
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

grade

analytical standard

product line

PESTANAL®

form

neat

CofA

certificate is enclosed in each package

shelf life

limited shelf life, expiry date on the label

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Agriculture
Environmental

format

neat

SMILES string

ClC1=CC=C(S(CC2=CC=C(Cl)C=C2)(=O)=O)C=C1

InChI

1S/C13H10Cl2O2S/c14-11-3-1-10(2-4-11)9-18(16,17)13-7-5-12(15)6-8-13/h1-8H,9H2

InChI key

OWFDXWCBYKXLRB-UHFFFAOYSA-N

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Codes

Xi

Risk Statement

36/38

Safety Statement

26

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.