35831

Supelco

1,2-Dichloro-4-nitrobenzene

PESTANAL®, analytical standard

Synonym(s):
3,4-Dichloronitrobenzene, asym.-Nitro-o-dichlorobenzene
Linear Formula:
Cl2C6H3NO2
CAS Number:
Molecular Weight:
192.00
Beilstein/REAXYS Number:
1818163
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

form

neat

shelf life

limited shelf life, expiry date on the label

application(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

255-256 °C (lit.)

mp

39-41 °C (lit.)

Featured Industry

Agriculture
Environmental

format

neat

SMILES string

[O-][N+](=O)c1ccc(Cl)c(Cl)c1

InChI

1S/C6H3Cl2NO2/c7-5-2-1-4(9(10)11)3-6(5)8/h1-3H

InChI key

NTBYINQTYWZXLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Flash Point(F)

255.2 °F

Flash Point(C)

124 °C

Certificate of Analysis

Certificate of Origin

Shingo Arakawa et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(9), 1545-1552 (2010-06-22)
A specific substrate to Mu class glutathione S-transferase (GST), 1,2-dichloro-4-nitrobenzene (DCNB), was administered to mice with a disrupted GST Mu 1 gene (Gstm1-null mice) to investigate the in vivo role of murine Gstm1 in toxicological responses to DCNB. A single...
Toshiyuki Watanabe et al.
Archives of toxicology, 78(4), 218-225 (2003-12-20)
Liver and kidney glutathione S-transferase (GST) activities to 1,2-dichloro-4-nitrobenzene (DCNB) as a substrate (GST-D activities) were measured in 280 dogs from five different breeders, and significant individual differences in this activity were observed in both organs. Interestingly, 34 out of...
V R Yoxall et al.
Archives of toxicology, 78(8), 477-482 (2004-03-23)
Rats were exposed to black tea (2.5% w/v) as their sole drinking liquid for either 1 day or 1 month, while controls were maintained on water. After this treatment period, all animals received a single oral dose IQ (2-amino-3-methylimidazo-[4,5-f]quinoline), and...
D A Basketter et al.
Contact dermatitis, 34(1), 55-58 (1996-01-01)
Unlike the closely related chemical dinitrochlorobenzene (DNCB), which is a very strong contact allergen, dichloronitrobenzene (DCNB) has been widely regarded as a non-allergen and, as such, a useful control for its strongly sensitizing counterpart. Nevertheless, it is still an organic...
V A Catania et al.
Life sciences, 68(4), 467-474 (2001-02-24)
Gender-related differences and the regulation by testosterone of glutathione S-transferase were studied in rat jejunum. We analyzed enzyme activity and the relative content of GST subunits. Four experimental groups of adult rats were studied: normal males, castrated males, castrated males...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.