Skip to Content
MilliporeSigma

370096

Oxazine 1 perchlorate

Dye content 85 %

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H26ClN3O5
CAS Number:
Molecular Weight:
423.89
UNSPSC Code:
12352200
EC Number:
246-465-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


composition

Dye content, 85%

mp

198-203 °C (lit.)

λmax

644 nm

SMILES string

[O-]Cl(=O)(=O)=O.CCN(CC)c1ccc2N=C3C=C\C(C=C3Oc2c1)=[N+](\CC)CC





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library