Documents

37440

Sigma-Aldrich

Dihydroxyacetone phosphate dilithium salt

~95% (enzymatic)

Synonym(s):
1,3-Dihydroxy-2-propanone 1-phosphate dilithium salt, DHAP, 1-Hydroxy-3-(phosphonooxy)-2-propanone dilithium salt
Empirical Formula (Hill Notation):
C3H5Li2O6P
CAS Number:
Molecular Weight:
181.92
Beilstein/REAXYS Number:
1708891
MDL number:
NACRES:
NA.25

assay

~95% (enzymatic)

impurities

≤0.2% D-glyceraldehyde-3-phosphate
≤2% solvent

solubility

H2O: 10 mg/mL, clear, faintly yellow

storage temp.

−20°C

SMILES string

[Li+].[Li+].OCC(=O)COP([O-])([O-])=O

InChI

1S/C3H7O6P.2Li/c4-1-3(5)2-9-10(6,7)8;;/h4H,1-2H2,(H2,6,7,8);;/q;2*+1/p-2

InChI key

QWIKESRFRWLYIA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dihydroxyacetone phosphate (DHAP) is a metabolic intermediate involved in many pathways, including glycolysis, gluconeogenesis, glycerol metabolism, phosphatidic acid synthesis, fat metabolism, and the Calvin cycle.

Other Notes

Substrate for the assay of triosephosphate isomerase

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis

Certificate of Origin

Triosephosphate isomerase from human erythrocytes.
R W Gracy
Methods in enzymology, 41, 442-447 (1975-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.