39937

SAFC

Methyl laurate

PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.

Synonym(s):
Methyl dodecanoate, Lauric acid methyl ester, Methyl laurate
Linear Formula:
CH3(CH2)10CO2CH3
CAS Number:
Molecular Weight:
214.34
Beilstein/REAXYS Number:
1767780
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.21

Quality Level

biological source

synthetic

Agency/Method

non-compendial

product line

PharmaGrade

assay

≥99.0% (rel.)
98.0-102.0% (w/w) (GC)

form

liquid

impurities

≤3000 ppm residual solvent (methanol) (GC)

refractive index

n20/D 1.429-1.433
n20/D 1.431 (lit.)

bp

262 °C/766 mmHg (lit.)

mp

4-5 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤500 ppm

cation traces

As: ≤2 ppm
Cd: ≤2 ppm
Cr: ≤2 ppm
Fe: ≤50 ppm
Hg: ≤2 ppm
Ni: ≤2 ppm
Pb: ≤2 ppm

suitability

corresponds for H-NMR

SMILES string

CCCCCCCCCCCC(=O)OC

InChI

1S/C13H26O2/c1-3-4-5-6-7-8-9-10-11-12-13(14)15-2/h3-12H2,1-2H3

InChI key

UQDUPQYQJKYHQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PharmaGrade products are manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. For more information on the PharmaGrade Program, visit sigma-aldrich.com/safc-pharmagrade.

Analysis Note

impurities (GC, area%, for information): methyl dodecanoate, methyl undecanoate, dodecanoic acid, methyl tridecanoate, methyl myristate, each unknown impurity

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

RIDADR

UN 3082 9 / PGIII

WGK Germany

WGK 1

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup

D A Dimas et al.
AAPS PharmSciTech, 1(2), E16-E16 (2004-01-20)
The effects of coating thickness, type of adhesive, and type and concentration of enhancer on the mechanical properties of two acrylic pressure-sensitive adhesives (PSAs) were investigated using a 2(4) factorial design and an optimization technique. Sixteen formulations containing 0% or...
E Barboza-Silva et al.
Oral microbiology and immunology, 24(4), 265-271 (2009-07-04)
The arginine deiminase system (ADS) of oral bacteria is a major generator of alkali (ammonia) in dental plaque and is considered to have anticaries effects. However, many of the antimicrobial agents used in oral care products may reduce alkali production...
Y Komata et al.
Biological & pharmaceutical bulletin, 17(5), 705-708 (1994-05-01)
The effect of long chain fatty acid (FA) and its analogs on the accumulation of thiamine disulfide (TDS) in rat skin using propylene glycol as a vehicle was studied in vitro. Lauric acid (12:0) increased the accumulation of TDS in...
Néstor M Carballeira et al.
Lipids, 40(10), 1063-1068 (2005-12-31)
The hitherto unknown 2-methylsulfanyldecanoic acid and 2-methylsulfanyldodecanoic acid were synthesized from methyl decanoate and methyl dodecanoate, respectively, through the reaction of lithium diisopropylamide and dimethyldisulfide in THF followed by saponification with potassium hydroxide in ethanol. Both alpha-methylsulfanylated FA were cytotoxic...
Jitender Sharma et al.
Journal of bioscience and bioengineering, 100(6), 662-666 (2006-02-14)
Efficient selective synthesis of the secondary amide surfactant N-methyl lauroylethanolamide from methyl laurate and N-methylethanol amine by carrier-fixed Chirazyme L-2 (Candida antarctica) using a kinetic strategy has been demonstrated. When different solvents were screened for product yields using Chirazyme L-2...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.