MilliporeSigma
All Photos(1)

41866

Supelco

(S)-(+)-7,8-Dihydrokavain

analytical standard

Synonym(s):
(6S)-5,6-Dihydro-4-methoxy-6-(2-phenylethyl)-2H-pyran-2-one, 7,8-Dihydrokawain, Dihydrokavain, Marindinin
Empirical Formula (Hill Notation):
C14H16O3
CAS Number:
Molecular Weight:
232.28
Beilstein:
15362
MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

optical activity

[α]/D 29±2°, c = 0.5 in methanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COC1=CC(=O)O[C@@H](CCc2ccccc2)C1

InChI

1S/C14H16O3/c1-16-13-9-12(17-14(15)10-13)8-7-11-5-3-2-4-6-11/h2-6,10,12H,7-9H2,1H3/t12-/m0/s1

InChI key

VOOYTQRREPYRIW-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service