All Photos(1)

41878

Supelco

Pentadecanolide

analytical standard

Synonym(s):
1-Oxa-2-cyclohexadecanone, 15-Pentadecanolide, Pentalide, 15-Hydroxypentadecanoic acid lactone
Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
Beilstein:
143710
MDL number:

grade

analytical standard

Quality Level

assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

bp

137 °C/2 mmHg (lit.)

mp

34-38 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

Featured Industry

Cleaning Products
Cosmetics
Food and Beverages
Personal Care

SMILES string

O=C1CCCCCCCCCCCCCCO1

InChI

1S/C15H28O2/c16-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-17-15/h1-14H2

InChI key

FKUPPRZPSYCDRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentadecanolide is one of the major components, isolated from the root of Angelica archangelica L., species. It is a synthetic musk, which exists as a widespread contaminant in the environment.

Application

Pentadecanolide may be used as an analytical reference standard for the quantification of the analyte in fish muscles and environmental samples using chromatography techniques.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Certificate of Analysis

Certificate of Origin

Study of Organic Pollutants in the Muscles of Fish Collected From El-mahmodia Stream At El-beheira Governorate, Egypt
Radwan HE, et al.
International Journal of Aquaculture Research and Development, 1(1), 19-19 (2019)
Nicola R Sumner et al.
Environmental pollution (Barking, Essex : 1987), 158(1), 215-222 (2009-08-07)
Synthetic musks are ubiquitous contaminants in the environment. Compartmental distributions (dissolved, suspended particle associated and sedimentary) of the compounds throughout an axial estuarine transect and in coastal waters are reported. High concentrations of Galaxolide (HHCB) and Tonalide (AHTN) (987-2098 ng/L...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service