MilliporeSigma
All Photos(1)

42688

Supelco

Oxypurinol

analytical standard

Empirical Formula (Hill Notation):
C5H4N4O2
CAS Number:
Molecular Weight:
152.11
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O=C1NC(=O)c2cn[nH]c2N1

InChI

1S/C5H4N4O2/c10-4-2-1-6-9-3(2)7-5(11)8-4/h1H,(H3,6,7,8,9,10,11)

InChI key

HXNFUBHNUDHIGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

An allopurinol metabolite.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Drugs in R&D, 5(3), 171-175 (2004-05-14)
Oxipurinol [alloxanthine, Oxyprim, oxypurinol] is the active metabolite of the only commercially available xanthine oxidase inhibitor, allopurinol. Oxipurinol is also a xanthine oxidase inhibitor. Oxipurinol is currently being developed by Cardiome Pharma. It is waiting for approval in the US
Richard O Day et al.
Clinical pharmacokinetics, 46(8), 623-644 (2007-07-28)
Allopurinol is the drug most widely used to lower the blood concentrations of urate and, therefore, to decrease the number of repeated attacks of gout. Allopurinol is rapidly and extensively metabolised to oxypurinol (oxipurinol), and the hypouricaemic efficacy of allopurinol
James Yun et al.
Journal of immunology (Baltimore, Md. : 1950), 192(7), 2984-2993 (2014-03-05)
Allopurinol (ALP) hypersensitivity is a major cause of severe cutaneous adverse reactions and is strongly associated with the HLA-B*58:01 allele. However, it can occur in the absence of this allele with identical clinical manifestations. The immune mechanism of ALP-induced severe
C C Wen et al.
Clinical pharmacology and therapeutics, 97(5), 518-525 (2015-02-14)
The first-line treatment of hyperuricemia, which causes gout, is allopurinol. The allopurinol response is highly variable, with many users failing to achieve target serum uric acid (SUA) levels. No genome-wide association study (GWAS) has examined the genetic factors affecting allopurinol
Lisa K Stamp et al.
Rheumatology (Oxford, England), 53(11), 1958-1965 (2014-06-06)
The aims of this study were to establish whether, in patients with gout, MPO is released from neutrophils and urate is oxidized to allantoin and if these effects are attenuated by allopurinol. MPO, urate, allantoin and oxypurinol were measured in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service