MilliporeSigma
All Photos(1)

428965

Sigma-Aldrich

Sodium hydrogen sulfate

ACS reagent, chunks

Synonym(s):
Sodium hydrogen sulfate, fused
Linear Formula:
Na2S2O7 · NaHSO4
PubChem Substance ID:

grade

ACS reagent

form

chunks

composition

acidity, 39.0-42.0% (as H2SO4)

impurities

≤0.01% NH4OH ppt.
≤0.01% insolubles

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

Ca: ≤0.002%
Fe: ≤0.002%
Mg: ≤0.001%
heavy metals (as Pb): ≤5 ppm

SMILES string

[Na+].[Na+].[Na+].OS([O-])(=O)=O.[O-]S(=O)(=O)OS([O-])(=O)=O

Components

Mixture of sodium pyrosulfate, Na2S2O7 and sodium hydrogen sulfate, NaHSO4.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service