All Photos(1)

43341

Supelco

α,α-Dimethylphenethyl acetate

analytical standard

Empirical Formula (Hill Notation):
C12H16O2
CAS Number:
Molecular Weight:
192.25
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water (calc. on elemental analysis)

refractive index

n20/D 1.4925 (lit.)

bp

250 °C (lit.)

density

0.998 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)OC(C)(C)Cc1ccccc1

InChI

1S/C12H16O2/c1-10(13)14-12(2,3)9-11-7-5-4-6-8-11/h4-8H,9H2,1-3H3

InChI key

FLUWAIIVLCVEKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service