All Photos(1)

46552

Supelco

β-Estradiol 3-benzoate

VETRANAL®, analytical standard

Synonym(s):
3-Benzoyloxy-17β-estrol, 1,3,5(10)-Estratriene-3,17β-diol 3-benzoate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3-benzoate
Empirical Formula (Hill Notation):
C25H28O3
CAS Number:
Molecular Weight:
376.49
Beilstein:
3107526
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

191-198 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])CCc4cc(OC(=O)c5ccccc5)ccc24

InChI

1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23+,25+/m1/s1

InChI key

UYIFTLBWAOGQBI-BZDYCCQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes.

Legal Information

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Chandra Suma J Miryala et al.
The journal of sexual medicine, 10(2), 350-361 (2012-11-01)
The selective serotonin reuptake inhibitor (SSRI), fluoxetine, leads to sexual dysfunction in a substantial proportion of women. In studies with the Fischer inbred rat, the 5-HT(1A) receptor has been implicated in this sexual dysfunction. Whether this association with 5-HT(1A) receptors
C M Martins et al.
Theriogenology, 78(5), 974-980 (2012-07-24)
Two experiments evaluated the effects of timing of the induction of ovulation in superstimulated lactating Holstein donor cows that were fixed-time artificially inseminated. Secondary objectives were to evaluate the effects of the timing of progesterone (P4) device removal (Experiment 1)
Shilpa Bharti et al.
Fertility and sterility, 99(1), 140-148 (2012-09-25)
To investigate the antiestrogenic effect of clomiphene citrate (CC) in male rats estrogenized with estradiol-3-benzoate (EB). Prospective experimental study. Laboratory. Adult male albino rats (Holtzman strain). CC was given alone or in combination with EB. Testicular function and steroidogenic enzyme
S A A Butler et al.
Reproduction in domestic animals = Zuchthygiene, 47(3), 463-471 (2012-04-14)
The primary objective of this study was to investigate the impact of animal-level factors including energy balance and environmental/management stress, on the ovarian function of Bos indicus heifers treated to synchronize ovulation. Two-year-old Brahman (BN) (n = 30) and BN-cross
S A A Edwards et al.
Animal reproduction science, 137(3-4), 129-136 (2013-02-05)
The aim of this study was to investigate the effects on follicle stimulating hormone (FSH) secretion and dominant follicle (DF) growth, of treatment of Bos indicus heifers with different combinations of intra-vaginal progesterone releasing devices (IPRD), oestradiol benzoate (ODB), PGF2α

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service