MilliporeSigma
All Photos(1)

49522

Millipore

m-Lauryl Sulfate Broth, Vegitone

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
Vegitone m-Lauryl Sulfate Broth
NACRES:
NA.85

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

lactose, 30.0 g/L
peptone (vegetable), 39.0 g/L
phenol red, 0.2 g/L
sodium lauryl sulfate, 1.0 g/L
yeast extract, 6.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective for Enterobacter spp.
selective for Escherichia coli
selective for Salmonella spp.
selective for coliforms

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
173494083489916
Lauryl sulfate Broth suitable for microbiology, NutriSelect® Plus

Millipore

17349

Lauryl sulfate Broth

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

composition

lactose, 30.0 g/L , peptone (vegetable), 39.0 g/L , phenol red, 0.2 g/L , sodium lauryl sulfate, 1.0 g/L , yeast extract, 6.0 g/L

composition

dipotassium hydrogen phosphate, 2.75 g/L , lactose, 5 g/L , potassium dihydrogen phosphate, 2.75 g/L , sodium chloride, 5 g/L , sodium lauryl sulfate, 0.1 g/L , tryptose, 20 g/L

composition

ammonium sulfate, 1 g/L , calcium chloride, 0.02 g/L , L-cysteine hydrochloride, 1 g/L , dextrose, 5 g/L , magnesium sulfate, 0.2 g/L , monopotassium phosphate, 15 g/L , sodium chloride, 5 g/L , starch, soluble, 1 g/L , tryptone (vegetable), 4 g/L , tryptose (vegetable), 10 g/L , vegetable infusion powder, 10 g/L , yeast extract, 5 g/L

composition

dipotassium phosphate, 2.75 g/L , enzymatic digest of plants and animal tissue, 20 g/L , lactose, 5 g/L , lauryl sulfate, 0.1 g/L , monopotassium phosphate, 2.75 g/L , sodium chloride, 34 g/L

manufacturer/tradename

NutriSelect® Plus

manufacturer/tradename

NutriSelect® Plus

manufacturer/tradename

NutriSelect® Plus

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

final pH

6.8±0.2 (25 °C)

final pH

7.2±0.2 (25 °C)

final pH

6.8±0.2 (25 °C)

Application

A medium where the animal derived peptone was replaced by a plant peptone. It is used for enumeration of Escherichia coli and coliforms in water, using membrane filter technique. It is recommended by ISO Committee under the specification ISO 9308-1:1990.

Preparation Note

Suspend 76.2 g in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Dispense as desired and sterilize by steaming for 30 minutes on three consecutive days or by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service