53156

Supelco

Stearoyl-L-carnitine-(N-methyl-d3)

analytical standard

Synonym(s):
Octadecanoyl-L-carnitine-(N-methyl-d3), L-Carnitine-(methyl-d3) octadecanoyl ester, (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxooctadecyl)oxy-1-propanaminium inner salt
Empirical Formula (Hill Notation):
C25D3H46NO4
CAS Number:
Molecular Weight:
430.68
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

assay

≥97.0% (TLC)

optical activity

α/D -13.0±2.0°, c = 0.1 in methanol

CofA

certificate is enclosed in each package

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99.0%

format

neat

storage temp.

−20°C

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.