All Photos(1)

54364

Supelco

N-Stearoylglycine

analytical standard

Synonym(s):
NSC 15863, N-(1-Oxooctadecyl)glycine, 2-Octadecanamidoacetic acid, 2-(Octadecanoylamino)acetic acid, N-Octadecanoylglycine
Empirical Formula (Hill Notation):
C20H39NO3
CAS Number:
Molecular Weight:
341.53
Beilstein:
1801426
MDL number:

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(CNC(CCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C20H39NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19(22)21-18-20(23)24/h2-18H2,1H3,(H,21,22)(H,23,24)

InChI key

UEYROBDNFIWNST-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service