MilliporeSigma
All Photos(1)

56931

Supelco

2,6-Diisopropylphenol

analytical standard

Synonym(s):
2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, Propofol
Linear Formula:
[(CH3)2CH]2C6H3OH
CAS Number:
Molecular Weight:
178.27
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

Quality Level

vapor pressure

5.6 mmHg ( 100 °C)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.512-1.516
n20/D 1.514 (lit.)

bp

256 °C/764 mmHg (lit.)

mp

18 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)c1cccc(C(C)C)c1O

InChI

1S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13H,1-4H3

InChI key

OLBCVFGFOZPWHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

1 of 4

This Item
D126608P-076Y0000016
2,6-Diisopropylphenol analytical standard

Supelco

56931

2,6-Diisopropylphenol

Propofol solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Supelco

P-076

Propofol solution

Propofol European Pharmacopoeia (EP) Reference Standard

Y0000016

Propofol

grade

analytical standard

grade

-

grade

certified reference material

grade

pharmaceutical primary standard

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

-

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

-

application(s)

forensics and toxicology

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

single component solution

format

neat

vapor pressure

5.6 mmHg ( 100 °C)

vapor pressure

5.6 mmHg ( 100 °C)

vapor pressure

-

vapor pressure

5.6 mmHg ( 100 °C)

assay

≥98.0% (GC)

assay

97%

assay

-

assay

-

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service