MilliporeSigma
All Photos(1)

60467

Sigma-Aldrich

Potassium permanganate

tested according to Ph. Eur.

Synonym(s):
Kalii permanganas
Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
EC Number:
MDL number:
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

assay

99.0-100.5%

form

crystalline

reaction suitability

reagent type: oxidant

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Suitable for heterogeneous oxidations.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Arnaud Beduneau et al.
PloS one, 4(2), e4343-e4343 (2009-02-03)
We posit that the same mononuclear phagocytes (MP) that serve as target cells and vehicles for a host of microbial infections can be used to improve diagnostics and drug delivery. We also theorize that physical and biological processes such as
Christopher H Gibbons et al.
Neurology, 72(17), 1479-1486 (2009-04-29)
To evaluate a novel method to quantify the density of nerve fibers innervating sweat glands in healthy control and diabetic subjects, to compare the results to an unbiased stereologic technique, and to identify the relationship to standardized physical examination and
F C Holstege et al.
Methods (San Diego, Calif.), 12(3), 203-211 (1997-07-01)
Open complex formation precedes initiation of transcription by RNA polymerases. In the analysis of transcription initiation from eukaryotic class II promoters, we have used promoter DNA structures that represent intermediates in open complex formation. We describe the preparation and isolation
Jessica M Terry et al.
Talanta, 99, 1051-1056 (2012-09-13)
We have explored the chemiluminescence response of amino acids and related compounds (including the tripeptide glutathione, and disulfides: cystine, homocystine and glutathione disulfide) with several new adaptations of the permanganate and tris(2,2'-bipyridine)ruthenium(III) ([Ru(bipy)(3)](3+)) reagents and a recently developed colloidal manganese(IV)
Matthias V Westphal et al.
ChemMedChem, 10(3), 461-469 (2015-01-30)
The tert-butyl group is a common motif in medicinal chemistry. Its incorporation into bioactive compounds is often accompanied by unwanted property modulation, such as increased lipophilicity and decreased metabolic stability. Several alternative substituents are available for the drug discovery process.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service