61370

Supelco

(−)-Carveol, mixture of isomers

analytical standard

Synonym(s):
p-Mentha-6,8-dien-2-ol
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

grade

analytical standard

assay

≥98% (sum of isomers, GC)

form

neat

CofA

certificate is enclosed in each package

shelf life

limited shelf life, expiry date on the label

application(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.496 (lit.)

bp

226-227 °C/751 mmHg (lit.)

density

0.958 g/mL at 25 °C (lit.)

Featured Industry

Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)[C@@H]1CC=C(C)C(O)C1

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9-11H,1,5-6H2,2-3H3/t9-,10?/m1/s1

InChI key

BAVONGHXFVOKBV-YHMJZVADSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Target Organs

Respiratory system

Hazard Codes

Xi

Risk Statement

36/37/38

Safety Statement

26-37/39

RIDADR

NONH for all modes of transport

WGK Germany

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Jenna L E Morrish et al.
Biotechnology and bioengineering, 101(4), 768-775 (2008-05-15)
The microbial biotransformation of (-)-trans-carveol to the flavor and fragrance compound (R)-(-)-carvone by Rhodococcus erythropolis DCL14 was carried out in a 3 L two phase partitioning bioreactor with an immiscible liquid second phase in an effort to improve upon the...
Jenna L E Morrish et al.
Biotechnology letters, 30(7), 1245-1250 (2008-03-11)
The aqueous substrate and product toxicity thresholds in the microbial biotransformation of (-)-trans-carveol to the fragrance/flavor compound (R)-(-)-carvone by Rhodococcus erythropolis were determined. Above aqueous phase concentrations of approx. 500 mg carveol/l and 200-600 mg carvone/l, the biotransformation activity of...
Aleem U Khand et al.
Journal of the American College of Cardiology, 42(11), 1944-1951 (2003-12-10)
This study examined the relative merits of digoxin, carvedilol, and their combination for the management of patients with atrial fibrillation (AF) and heart failure (HF). In patients with AF and HF, both digoxin and beta-blockers reduce the ventricular rate, and...
Alison Li Lin et al.
Life sciences, 79(1), 21-29 (2006-01-27)
2,6-Diisopropylphenol (Propofol) is a short-acting intravenous anesthetic that is rapidly metabolized by glucuronidation and ring hydroxylation catalyzed by cytochrome P450. The goal of this research was to determine whether dietary monoterpene alcohols (MAs) could be used to prolong the anesthetic...
S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S88-S90 (2008-07-22)
A toxicologic and dermatologic review of laevo-carveol when used as a fragrance ingredient is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.