All Photos(1)

667390

Supelco

Methanol solution

contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC

Empirical Formula (Hill Notation):
CH4O
CAS Number:
Molecular Weight:
32.04
Beilstein:
3662861
MDL number:
PubChem Substance ID:

Quality Level

vapor density

1.11 (vs air)

vapor pressure

410 mmHg

form

liquid

contains

0.1 % (v/v) trifluoroacetic acid
5 % (v/v) water

expl. lim.

5.5-44 % (lit.)

application(s)

HPLC: suitable

bp

65.4 °C (lit.)

mp

-98 °C (lit.)

density

0.807 g/mL at 25 °C

UV absorption

λ: 210 nm Amax: ≤1.30
λ: 230 nm Amax: ≤0.50
λ: 254 nm Amax: ≤0.05

Featured Industry

Food and Beverages

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

4×4 L in glass bottle

Preparation Note

Product filtered through a 0.2 μm filter

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK Germany

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Certificate of Analysis

Certificate of Origin

Optimum concentration of trifluoroacetic acid for reversed-phase liquid chromatography of peptides revisited.
Chen Y, et al.
Journal of Chromatography A, 1043(1), 9-18 (2004)
Laurence C Cadalbert et al.
PloS one, 10(6), e0131066-e0131066 (2015-06-27)
Barth syndrome is an X-linked mitochondrial disease, symptoms of which include neutropenia and cardiac myopathy. These symptoms are the most significant clinical consequences of a disease, which is increasingly recognised to have a variable presentation. Mutation in the Taz gene...
Mark Barszczyk et al.
Acta neuropathologica, 128(6), 863-877 (2014-08-15)
Pediatric ependymomas are highly recurrent tumors resistant to conventional chemotherapy. Telomerase, a ribonucleoprotein critical in permitting limitless replication, has been found to be critically important for the maintenance of tumor-initiating cells (TICs). These TICs are chemoresistant, repopulate the tumor from...
Erik Müllers et al.
Cell cycle (Georgetown, Tex.), 13(17), 2733-2743 (2014-12-09)
Upon DNA damage, cell cycle progression is temporally blocked to avoid propagation of mutations. While transformed cells largely maintain the competence to recover from a cell cycle arrest, untransformed cells past the G1/S transition lose mitotic inducers, and thus the...
Ning Liang et al.
The Journal of experimental medicine, 211(11), 2249-2263 (2014-10-08)
Genetic studies have shown that the tuberous sclerosis complex (TSC) 1-TSC2-mammalian target of Rapamycin (mTOR) and the Hippo-Yes-associated protein 1 (YAP) pathways are master regulators of organ size, which are often involved in tumorigenesis. The crosstalk between these signal transduction...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service