All Photos(1)

68225

Supelco

1-Octen-3-ol

analytical standard

Synonym(s):
Pentyl vinyl carbinol
Linear Formula:
CH3(CH2)4CH(OH)CH=CH2
CAS Number:
Molecular Weight:
128.21
Beilstein:
1744110
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.437 (lit.)
n20/D 1.437

bp

84-85 °C/25 mmHg (lit.)

density

0.837 g/mL at 20 °C
0.83 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCC(O)C=C

InChI

1S/C8H16O/c1-3-5-6-7-8(9)4-2/h4,8-9H,2-3,5-7H2,1H3

InChI key

VSMOENVRRABVKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Certificate of Analysis

Certificate of Origin

Giulianna Echeverria Macedo et al.
Ecotoxicology and environmental safety, 206, 111232-111232 (2020-09-06)
Fungal volatile organic compounds (VOCs) comprise a group of compounds commonly found in damp or water-damaged indoor places affecting air quality. Indoor fungal pollution is a severe threat to human health, contributing to the onset of allergic diseases. The compound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service