MilliporeSigma
All Photos(1)

73044BC

Millipore

GDS Sample Wells

BioControl, For use with GDS Sample Wells Base

Synonym(s):
Sample Wells for PCR Sample Prep

manufacturer/tradename

BioControl Systems 73044

packaging

pkg of (Bag of 200)

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

General description

GDS Sample Wells are the disposables required to perform Assurance® GDS assays. Food sample is added to the sample wells, along with a Concentration Reagent (IMS beads) as part of the initial PCR sample preparation for GDS. Adhesive film strips are used to seal the sample wells.

Application

GDS Sample Wells are the disposables used during PCR sample preparation for Assurance® GDS assays. The sample wells are to be used with the GDS Sample Wells Base.

Components

Bag of 200

Compatibility

BioControl GDS

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

Assurance is a registered trademark of BioControl Systems, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service