All Photos(1)

74002

Supelco

2-Methylpiperidine

analytical standard

Synonym(s):
α-Pipecoline, NSC 31047, 2-Pipecoline, NSC 462
Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
Beilstein:
79804
EC Number:
MDL number:
PubChem Substance ID:

grade

analytical standard

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤2.5% water

refractive index

n20/D 1.445 (lit.)

bp

118-119 °C/753 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

SMILES string

CC1CCCCN1

InChI

1S/C6H13N/c1-6-4-2-3-5-7-6/h6-7H,2-5H2,1H3

InChI key

NNWUEBIEOFQMSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service