78000BC

Millipore

Gemini

BioControl, ELISA Automation

manufacturer/tradename

BioControl Systems 78000
Gemini®

application(s)

ELISA: suitable
pathogen testing: suitable

Featured Industry

Food and Beverages

General description

Gemini® is a fully automated microplate processor that is compatible with all EIA assays and is pre-programmed for BioControl′s TRANSIA Plate assays. GEMINI offers increased capacity over other systems for high throughput testing.

Features and Benefits

  • Full Automation of EIA Procedure
  • Versatility to Meet Lab Demands
  • Unique Modular Design

Testing Method

ELISA

Legal Information

Gemini is a registered trademark of Stratec

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Related Content
TRANSIA AG (Enzyme ImmunoAssay) and TRANSIA® PLATE incorporate proven antibody-antigen "sandwich" assay technology for the detection of food borne pathogens in both product and environmental samples
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.