MilliporeSigma
All Photos(1)

81966

Supelco

Devarda′s alloy

for preparative purposes, grit

Linear Formula:
AlCuZn
CAS Number:
MDL number:
PubChem Substance ID:

grade

for preparative purposes

form

grit

composition

aluminum, ~45%
copper, ~50%
zinc, ~5%

mp

550 °C (lit.)

application(s)

forensics and toxicology

SMILES string

[Al].[Cu].[Zn]

InChI

1S/Al.Cu.Zn

InChI key

MUBKMWFYVHYZAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service