MilliporeSigma
All Photos(1)

83700

Supelco

Rhodanine

for spectrophotometric det. of gallic acid, ≥99.0%

Synonym(s):
2-Thioxo-4-thiazolidinone
Empirical Formula (Hill Notation):
C3H3NOS2
CAS Number:
Molecular Weight:
133.19
Beilstein:
110701
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.21

Quality Level

assay

≥99.0% (HPLC)
≥99.0%

form

crystals

quality

for spectrophotometric det. of gallic acid

technique(s)

UV/Vis spectroscopy: suitable

mp

165-169 °C (lit.)
165-169 °C

solubility

methanol: 0.5 g/10 mL, clear, colorless to greenish-yellow

SMILES string

O=C1CSC(=S)N1

InChI

1S/C3H3NOS2/c5-2-1-7-3(6)4-2/h1H2,(H,4,5,6)

InChI key

KIWUVOGUEXMXSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rhodanine is a drug, which prevents the multiplication of echovirus 12.

Other Notes

Reagent for the gravimetric determination of silver; for the photometric determination of gallic acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service