90059

Supelco

N-3-(2-Benzothiazolyl)-4-(tert-butyldiphenylsilyloxy)phenylbenzamide

for colorimetric determination of fluoride

Empirical Formula (Hill Notation):
C36H32N2O2SSi
CAS Number:
Molecular Weight:
584.80
MDL number:
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

assay

≥95.0% (HPLC)

application(s)

UV/Vis spectroscopy: suitable

SMILES string

CC(C)(C)Si(Oc1ccc(NC(=O)c2ccccc2)cc1-c3nc4ccccc4s3)(c5ccccc5)c6ccccc6

InChI

1S/C36H32N2O2SSi/c1-36(2,3)42(28-17-9-5-10-18-28,29-19-11-6-12-20-29)40-32-24-23-27(37-34(39)26-15-7-4-8-16-26)25-30(32)35-38-31-21-13-14-22-33(31)41-35/h4-25H,1-3H3,(H,37,39)

InChI key

YFMSVSWQTBPEIP-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
A Rapid Aqueous Fluoride Ion Sensor with Dual Output Modes
Hu, R., et al.
Angewandte Chemie (International Edition in English), 49, 1-5 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.