Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

936677

Hoveyda-Grubbs Catalyst® M720 ChemBeads

greener alternative

solid, beads

Synonym(s):

(1,3-Bis-(2,4,6-trimethylphenyl)-2-imidazolidinylidene)dichloro(o-isopropoxyphenylmethylene)ruthenium ChemBeads, Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](2-isopropoxyphenylmethylene)ruthenium(II) ChemBeads, Grubbs Catalyst® C627 ChemBeads, Hoveyda-Grubbs Catalyst® 2nd Generation ChemBeads, Hoveyda-Grubbs Catalyst® M72 (C627) ChemBeads

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352100
NACRES:
NA.21

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Hoveyda-Grubbs Catalyst® M720 ChemBeads,

Quality Level

greener alternative product characteristics

Waste Prevention
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

product line

ChemBeads

form

beads, solid

composition

loading, 4-6 wt. %

reaction suitability

reagent type: catalyst

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Greener Chemistry. ChemBeads are advanced solid-coated beads for high-throughput experimentation in various chemical applications. They enable precise dispensing of solid reagents, allowing researchers to conduct reactions with minimal material use, thus supporting waste prevention and catalysis principles while ensuring accuracy and reproducibility. This technology enhances efficiency and streamlines experimental processes in chemistry. Click here for more information.
The ChemBeads product of second generation, phosphine-free Hoveyda-Grubbs Catalyst® M720. The catalyst has comparable reactivity to the first generation, but initiates more readily at lower temperatures. Efficient for metathesis of electron-deficient substrates including fluorinated olefins. Demonstrated in multiple large-scale pharmaceutical applications. Loaded on glass beads for use in high-throughput expermentation (HTE).

Application

Hoveyda-Grubbs Catalyst® M720 has be used as:
A catalyst for olefin cross-metathesis with fluorinated olefins (CM).[1]
A catalyst for ring-closing metathesis (RCM).[2]
A catalyst for ring-opening metathesis (ROM).[2]
A catalyst for a sequence of a metathesis reaction and subsequent dihydroxylation of the newly formed double bond.[3]
A ROM catalyst in mechanochemical polymer synthesis.[4]
A catalyst in the total synthesis of the Diterpenes (+)-Randainin D and (+)-Barekoxide.[4]
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For general uses, product is also available in powdered form (569755)

Legal Information

Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service