95334

Supelco

Glutaryl-L-carnitine-(N-methyl-d3) lithium salt

analytical standard

Empirical Formula (Hill Notation):
C12D3H18NO6 · xLi+
Molecular Weight:
278.32 (free acid basis)
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

optical activity

[α]/D -19.0 to -25.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99.0%

format

neat

storage temp.

−20°C

SMILES string

C[N+](C)(C([2H])([2H])[2H])C[C@H](OC(CCCC(O)=O)=O)CC([O-])=O.[Li]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Target Organs

Respiratory system

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.