MilliporeSigma
All Photos(1)

B1131

Sigma-Aldrich

Brilliant Blue G

Dye content ~60 % by elemental analysis

Synonym(s):
Coomassie Brilliant Blue G, Acid blue 90
Empirical Formula (Hill Notation):
C47H48N3NaO7S2
CAS Number:
Molecular Weight:
854.02
Colour Index Number:
42655
Beilstein:
5230822
EC Number:
MDL number:
PubChem Substance ID:

composition

Dye content, ~60% elemental analysis

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3C)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=CC(\C=C5C)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C47H49N3O7S2.Na/c1-6-49(31-35-11-9-13-43(29-35)58(51,52)53)40-21-25-45(33(4)27-40)47(37-15-17-38(18-16-37)48-39-19-23-42(24-20-39)57-8-3)46-26-22-41(28-34(46)5)50(7-2)32-36-12-10-14-44(30-36)59(54,55)56;/h9-30H,6-8,31-32H2,1-5H3,(H2,51,52,53,54,55,56);/q;+1/p-1

InChI key

RWVGQQGBQSJDQV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

P2X7 purinergic receptor antagonist.

Legal Information

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Søren U Nielsen et al.
The Journal of general virology, 89(Pt 10), 2507-2517 (2008-09-18)
Hepatitis C virus (HCV) particles found in vivo are heterogeneous in density and size, but their detailed characterization has been restricted by the low titre of HCV in human serum. Previously, our group has found that HCV circulates in blood
Valentina Murdica et al.
Fertility and sterility, 111(5), 897-908 (2019-04-29)
To characterize in depth and investigate the role of exosomes present in seminal plasma in affecting parameters underlying sperm activity. In vitro experimental study. Research hospital. Normozoospermic, severe asthenozoospermic, and post-vasectomy azoospermic men 18-55 years of age were considered for the study.
David E Pelayes et al.
Ophthalmic research, 48 Suppl 1, 21-25 (2012-08-29)
Brilliant blue G (BBG) is frequently used in chromovitrectomy to facilitate internal limiting membrane (ILM) peeling. A study was initiated to evaluate if heavy BBG is safe and effective in staining the ILM. We studied 30 eyes, 23 with idiopathic
Sungwon Han et al.
Analytical and bioanalytical chemistry, 405(6), 1821-1831 (2012-09-25)
Gel electrophoresis is routinely used to separate and analyse macromolecules in biological systems. Although many of these electrophoretic techniques necessitate the denaturing of the analytes prior to their analysis, blue native polyacrylamide gel electrophoresis (BN-PAGE) permits the investigation of proteins/enzymes
Song Wei et al.
Bioorganicheskaia khimiia, 38(5), 535-544 (2013-01-25)
A novel 4-(tetrahydro-2-furanmethoxy)-N-octadecyl-1,8-naphthalimide (4-TN1) blue emitting fluorophore was synthesized for solvent polarity probing and protein detection. The effect of various solvents on absorption and fluorescence spectra of 4-TNI was investigated. By comparison with other derivatives with heterocyclic electron-donating groups, 4-TNI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service