MilliporeSigma
All Photos(1)

B5133

Sigma-Aldrich

Brilliant Blue G

≥90% (TLC)

Synonym(s):
Coomassie Brilliant Blue G, Acid blue 90
Empirical Formula (Hill Notation):
C47H48N3NaO7S2
CAS Number:
Molecular Weight:
854.02
Colour Index Number:
42655
Beilstein:
5230822
MDL number:

assay

≥90% (TLC)

purified by

recrystallization

composition

Dye content, ≥90% elemental analysis

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3C)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=CC(\C=C5C)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C47H49N3O7S2.Na/c1-6-49(31-35-11-9-13-43(29-35)58(51,52)53)40-21-25-45(33(4)27-40)47(37-15-17-38(18-16-37)48-39-19-23-42(24-20-39)57-8-3)46-26-22-41(28-34(46)5)50(7-2)32-36-12-10-14-44(30-36)59(54,55)56;/h9-30H,6-8,31-32H2,1-5H3,(H2,51,52,53,54,55,56);/q;+1/p-1

InChI key

RWVGQQGBQSJDQV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

P2X7 purinergic receptor antagonist.

Preparation Note

Chromatographically purified

Other Notes

This product has been replaced by B0770-Sigma-Aldrich | Brilliant Blue G pure

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Søren U Nielsen et al.
The Journal of general virology, 89(Pt 10), 2507-2517 (2008-09-18)
Hepatitis C virus (HCV) particles found in vivo are heterogeneous in density and size, but their detailed characterization has been restricted by the low titre of HCV in human serum. Previously, our group has found that HCV circulates in blood
Kei Takayama et al.
Investigative ophthalmology & visual science, 53(11), 7389-7394 (2012-10-04)
We compared the phototoxicity of indocyanine green (ICG) and Brilliant Blue G (BBG) in cultured RPE cells under fluorescent lamp illumination imitating ambient light. Cultured human RPE line cells were stained with ICG or BBG solution at concentrations of clinical
Broc R Wenrich et al.
Analytical biochemistry, 428(2), 93-95 (2012-06-30)
The Bradford assay has been used reliably for decades to quantify protein in solution. The analyte is incubated in acidic solution of Coomassie Blue G-250 dye, during which reversible ionic and nonionic binding interactions form. Bradford assay color yields were
Sungwon Han et al.
Analytical and bioanalytical chemistry, 405(6), 1821-1831 (2012-09-25)
Gel electrophoresis is routinely used to separate and analyse macromolecules in biological systems. Although many of these electrophoretic techniques necessitate the denaturing of the analytes prior to their analysis, blue native polyacrylamide gel electrophoresis (BN-PAGE) permits the investigation of proteins/enzymes
Song Wei et al.
Bioorganicheskaia khimiia, 38(5), 535-544 (2013-01-25)
A novel 4-(tetrahydro-2-furanmethoxy)-N-octadecyl-1,8-naphthalimide (4-TN1) blue emitting fluorophore was synthesized for solvent polarity probing and protein detection. The effect of various solvents on absorption and fluorescence spectra of 4-TNI was investigated. By comparison with other derivatives with heterocyclic electron-donating groups, 4-TNI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service